Preparative LC columns
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
410910-101 410910-102 410910-301 410910-302 410910-501 410910-502 413910-101 413910-102 413910-301 413910-302 413910-501 413910-502 419910-301 419910-302 419910-501 419910-502 420212-901 420212-902 440905-901 440905-902 443905-101 443905-102 443905-901 443905-902 446905-101 446905-102 446905-301 446905-302 446905-901 446905-902 449905-101 449905-102 449905-301 449905-302 449905-501 449905-502 449905-901 449905-902 440910-901 440910-902 443910-901 443910-902 446910-102 449910-902
HEWL410910-101EA 3001 EUR InStock
HEWL410910-101 HEWL410910-102 HEWL410910-301 HEWL410910-302 HEWL410910-501 HEWL410910-502 HEWL413910-101 HEWL413910-102 HEWL413910-301 HEWL413910-302 HEWL413910-501 HEWL413910-502 HEWL419910-301 HEWL419910-302 HEWL419910-501 HEWL419910-502 HEWL420212-901 HEWL420212-902 HEWL440905-901 HEWL440905-902 HEWL443905-101 HEWL443905-102 HEWL443905-901 HEWL443905-902 HEWL446905-101 HEWL446905-102 HEWL446905-301 HEWL446905-302 HEWL446905-901 HEWL446905-902 HEWL449905-101 HEWL449905-102 HEWL449905-301 HEWL449905-302 HEWL449905-501 HEWL449905-502 HEWL449905-901 HEWL449905-902 HEWL440910-901 HEWL440910-902 HEWL443910-901 HEWL443910-902 HEWL446910-102 HEWL449910-902
Preparative LC columns
Chromatographiesäulen
These Prep LC columns are designed for high loadability to purify milligram to gram quantities of products.
  • High loadability for maximum sample purification
  • Easy scalability from 4,6 up to 50 mm ID for rapid method development
  • High throughput 21,2 mm ID cartridges for fast purification
  • Exceptional column stability and loadability up to pH 10
Prep 21,2 mm ID columns are available with preparative cartridge hardware. This reliable cartridge hardware makes it simple to use columns with different lengths to increase sample load. Guard columns are easily integrated onto these columns, providing superior protection of the analysis column.
Analytical size 4,6 mm ID scalar columns are available for method development and optimisation prior to scaling up to larger columns.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge