HPLC columns, Acclaim™ Trinity™ Q1
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
083244 079719
555-7384EA 411 EUR InStock
555-7384 555-7530
HPLC columns, Acclaim™ Trinity™ Q1
Chromatographiesäulen
The Acclaim™ Trinity™ Q1 column is a mixed-mode, silica-based application-specific column for high-resolution and high-throughput trace analysis of herbicides diquat and paraquat by LC-MS/MS and LC-UV methods.
  • No need for ion-pairing reagent use
  • LC/MS compatible
  • Direct injection possible with on-line SPE
  • Excellent resolution, good peak shape
Particle type: ultra pure, spherical silica
Bonded phase: mixed-mode reversed phase, anion exchange and cation exchange
pH range: 2,5 to 7,5
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge