HPLC columns, Pursuit™ XRs
Lieferant: VARIAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
A6001030X046 A6000030X020 A6000050X020 A6000050X046 A6000050X100 A6000050X212 A6000100X020 A6000100X030 A6000100X046 A6000100X212 A6000100X300 A6000150X020 A6000150X030 A6000150X040 A6000150X046 A6000150X100 A6000150X212 A6000250X020 A6000250X030 A6000250X040 A6000250X046 A6000250X100 A6000250X212 A6000250X300 A6001020X020 A6001050X020 A6001050X030 A6001050X046 A6001100X020 A6001100X030 A6001100X046 A6001150X020 A6001150X030 A6001150X046 A6001250X020 A6001250X046 A6002250X046 A6002250X100 A6002250X212 A6002250X500 A6004250X046 A6004250X212 A6004250X300 A6005050X020 A6005100X046 A6010100X046 A6010150X020 A6010150X040 A6010150X046 A6010250X040 A6010250X046 A6011050X020 A6011050X046 A6011100X020 A6011100X046 A6011150X020 A6020050X020 A6020050X046 A6020100X046 A6020150X020 A6020150X046 A6020250X020 A6020250X046 A6020250X100 A6020250X212 A6021030X020 A6021030X046 A6021050X020 A6021050X046 A6021100X020 A6021100X030 A6021100X046 A6021150X020 A6021150X030 A6021150X046 A6021250X020 A6021250X046 A6000150X300 A6002050X046S A6020100X212 A6022250X500
VARIA6001030X046EA 616 EUR InStock
VARIA6001030X046 VARIA6000030X020 VARIA6000050X020 VARIA6000050X046 VARIA6000050X100 VARIA6000050X212 VARIA6000100X020 VARIA6000100X030 VARIA6000100X046 VARIA6000100X212 VARIA6000100X300 VARIA6000150X020 VARIA6000150X030 VARIA6000150X040 VARIA6000150X046 VARIA6000150X100 VARIA6000150X212 VARIA6000250X020 VARIA6000250X030 VARIA6000250X040 VARIA6000250X046 VARIA6000250X100 VARIA6000250X212 VARIA6000250X300 VARIA6001020X020 VARIA6001050X020 VARIA6001050X030 VARIA6001050X046 VARIA6001100X020 VARIA6001100X030 VARIA6001100X046 VARIA6001150X020 VARIA6001150X030 VARIA6001150X046 VARIA6001250X020 VARIA6001250X046 VARIA6002250X046 VARIA6002250X100 VARIA6002250X212 VARIA6002250X500 VARIA6004250X046 VARIA6004250X212 VARIA6004250X300 VARIA6005050X020 VARIA6005100X046 VARIA6010100X046 VARIA6010150X020 VARIA6010150X040 VARIA6010150X046 VARIA6010250X040 VARIA6010250X046 VARIA6011050X020 VARIA6011050X046 VARIA6011100X020 VARIA6011100X046 VARIA6011150X020 VARIA6020050X020 VARIA6020050X046 VARIA6020100X046 VARIA6020150X020 VARIA6020150X046 VARIA6020250X020 VARIA6020250X046 VARIA6020250X100 VARIA6020250X212 VARIA6021030X020 VARIA6021030X046 VARIA6021050X020 VARIA6021050X046 VARIA6021100X020 VARIA6021100X030 VARIA6021100X046 VARIA6021150X020 VARIA6021150X030 VARIA6021150X046 VARIA6021250X020 VARIA6021250X046 VARIA6000150X300 VARIA6002050X046S VARIA6020100X212 VARIA6022250X500
HPLC columns, Pursuit™ XRs
Chromatographiesäulen
Pursuit XRs are ultra-purity silica based columns designed to increase productivity.
  • Maximum loadability
  • Excellent stability
  • Easy scalability
  • Used in analytical research and development, QC and preparative applications
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge