Shodex HILICpak VN-50 HILIC Column for Oligonucleotides and Oligosaccharides
Lieferant: Resonac
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
F6711400 F6711500 F7630500 F7630600 F6830100
SHOWF6711400EA 600 EUR InStock
SHOWF6711400 SHOWF6711500 SHOWF7630500 SHOWF7630600 SHOWF6830100
Shodex HILICpak VN-50 HILIC Column for Oligonucleotides and Oligosaccharides
Chromatographiesäulen
Robust polymer-based HILIC column for separation by size/chain length.
  • Polymer-based HILIC columns with diol (-OH) groups
  • Best for oligosaccharides and oligonucleotides
  • Suitable for oligomer separation which is not possible by SEC column or conventional HILIC columns
  • The modified diol groups on the packing material create the HILIC mode
  • Polymer-based packing material provides excellent chemical stability and minimum deterioration over an extended time period
  • pH stability: 2 to 13
  • Also suitable for evaporative light scattering detector (ELSD), corona charged aerosol detector (CAD), and mass spectrometry (LC-MS/MS)
The HILICpak VN-50 columns with diol groups are robust HILIC columns for separating analytes by their degree of polymerization. The polymeric base material polyvinyl alcohol offers great pH stability. The column housing is made of PEEK to reduce unwanted interaction.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge