Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ultrasonic Baths, Elmasonic Selects

Lieferant: ELMA SCHMIDBAUER

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

1106668 1106989 1106990 1106991 1106992 1106993 1106853
ELMA1106668EA 995 EUR InStock
ELMA1106668 ELMA1106989 ELMA1106990 ELMA1106991 ELMA1106992 ELMA1106993 ELMA1106853
Ultrasonic Baths, Elmasonic Selects
Ultraschallbäder

Powerful and individual - the versatile Elmasonic Select series for ultrasonic cleaning are equipped with 37 kHz ultrasonic frequency, five ultrasonic modes, the ability to save up to four cleaning programmes, a heating and tank volumes from 3 to 90 L (model Select 60 and larger have a drain).



  • Adjustable cleaning programmes

  • Memory for four individual cleaning programmes

  • Cleaning process in the ultrasonic cleaning device can be called up for reproducible results

  • Fast switching between favourite programmes

  • Auto-degas mode removes air bubbles in the cleaning liquid giving better cleaning results For temperature-sensitive parts or processes, the limit temperature can be set individually and the display changed to °C/°F


Select units feature five ultrasonic modes for best cleaning results and easy sample preparation:

Dynamic: Combines the ultrasonic modes pulse and sweep to increase the overall cleaning performance.

Pulse: Removes even stubborn contaminations through a pulsating sound field distribution.

Sweep: Uniform distribution of the ultrasonic power for most cleaning tasks.

Eco: For gentle cleaning with quiet operation and for a longer service life of the ultrasonic cleaning device.

Degas: Quick degassing of freshly prepared cleaning solutions and (HPLC) samples.



The models have a clear, easy-to-read display showing all relevant parameters eg. remaining cleaning time, set/actual temperature for precise adjustment. The user is always informed about the operating status of the device via clear symbols for eg. end of cleaning or over temperature as well as acoustic and visual feedback.



Lieferumfang:

Supplied with cover.



Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge