Oligomeric Amyloid-β (Aβ) ELISA Kit
Lieferant: BIOSENSIS PTY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BEK-2215-1P BEK-2215-2P
BSENBEK-2215-1PEA 398 EUR InStock
BSENBEK-2215-1P BSENBEK-2215-2P
Oligomeric Amyloid-β (Aβ) ELISA Kit
Tests ELISAs
The oligomeric form of Amyloid Beta peptide (Aβ, 1- 42) has been closely linked to Alzheimer’s Disease. Several ELISAs targeting Aβ have been developed; however, these ELISAs are known to cross-react with Amyloid Beta precursor protein (APP) and are poorly characterised against monomeric and oligomeric forms of the peptide.
The Biosensis MOAB-2 antibody, developed by LaDu and co-workers (Youmans K. et al., 2012), has been shown to specifically detect Aβ, but not the precursor molecule APP. When utilised in ELISAs, the oligomeric form of Aβ peptide (o-Aβ) can be assayed independently of the other forms of the molecule when assayed with the MOAB-2 monoclonal antibody.
The Biosensis oligomeric Aβ ELISA kit is a sandwich ELISA that allows the preferential quantification of oligomeric Aβ peptides. This kit is exclusive to Biosensis and consists of a pre-coated mouse monoclonal anti-Aβ capture antibody (MOAB-2), a biotinylated MOAB-2 detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of o-Aβ present in samples and protein standards.
The purpose of this kit is the in vitro qualitative measurement of oligomeric Aβ peptide levels in brain extracts and CSF samples from both transgenic mice and humans relative to a known, preformed o-Aβ standard for increased reproducibility. The inclusion of a highly validated oligomeric positive control sample results in a unique, ready-to-use ELISA kit.
Lieferumfang: Each kit contains 1 or 2× 96 pre-coated strip plates, protein standards, detection reagents, sample buffer and detailed instructions.
Achtung! Bitte beachten: For research use only. Not for diagnostic or clinical purposes.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge