Antibiotic assay discs, multodisc
Lieferant: LIOFILCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
95240 95250 95260 95290 95230
LIOF95240EA 204 EUR InStock
LIOF95240 LIOF95250 LIOF95260 LIOF95290 LIOF95230
Antibiotic assay discs, multodisc
Mikrobiologische Testsysteme Antibiotika-Tests
Multodisc with eight antimicrobial agents on a ring.
  • Each specially for a microorganism
Amikacin (AK), Ampicillin (AMP), Ampicillin-Sulbactam (AMS), Carbenicillin (CAR), Cefoxitin (FOX), Ceftazidime (CAZ), Cefuroxime (CXM), Chloramphenicol (C), Clindamycin (CD), Erythromycin (E), Gentamicin (CN), Levofloxacin (LEV), Linezolid (LNZ), Metronidazole (MTZ), Netilmicin (NET), Nitrofurantoin (F), Norfloxacin (NOR), Penicillin G (P), Piperacillin-Tazobactam (TZP), Rifampicin (RD), Streptomycin (S), Teicoplanin (TEC), Tetracycline (TE), Ticarcillin (TC), Tobramycin (TOB), Trimethoprim-Sulfamethoxazole (SXT), Vancomycin (VA). The numbers shows the concentration in μg.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge