Avian influenza A (H5N1) neuraminidase Influenza A Peptide (15 aa near C-terminus)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3423P
PRSI3423PEA 122 EUR InStock
PRSI3423P
Avian influenza A (H5N1) neuraminidase Influenza A Peptide (15 aa near C-terminus)
Proteine und Peptide
The peptide is used for blocking the activity of the avian influenza neuraminidase antibody.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge