Avian Influenza NS1 Influenza NS1 Peptide (14 aa near C-terminus)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3915P
PRSI3915PEA 122 EUR InStock
PRSI3915P
Avian Influenza NS1 Influenza NS1 Peptide (14 aa near C-terminus)
Proteine und Peptide
Avian influenza nonstructural protein 1 peptide is used for blocking the activity of avian influenza nonstructural protein 1 antibody.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge