Human recombinant IL15 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40-558
PRSI40-558EA 461 EUR InStock
PRSI40-558
Human recombinant IL15 (from E. coli)
Proteine und Peptide
IL-15 is an immunomodulating cytokine that stimulates the proliferation of T lymphocytes and shares many biological properties with IL-2. IL-15 exerts its biological activities primarily on T cells. It is also essential in the development, survival and activation of NK cells. Increased expression of IL-15 has been implicated with rheumatoid arthritis, inflammatory bowl disease and diseases affiliated with retroviruses HIV and HTLV-I. Human IL-15 is biologically active on mouse cells as measured by the dose-dependent stimulation of the proliferation of mouse CTLL cells.
  • Recombinant human (40-260) IL-15 is a 12.9 kDa protein consisting of 114 amino acid residues. Recombinant human IL-15 (40-558) is a 12.9 kDa protein consisting of 115 amino acid residues. Recombinant murine IL-15 is a 13.3 kDa protein consisting of 115 amino acid residues. Recombinant rat IL-15 is a 13.5 kDa protein consisting of 115 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge