H1N1 Neuraminidase peptide
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5245P
PRSI5245PEA 122 EUR InStock
PRSI5245P
H1N1 Neuraminidase peptide
Proteine und Peptide
H1N1 Neuraminidase peptide is meant to be used as a positive control in ELISA applications to detect either Seasonal or Swine-Origin Influenza A antibody.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge