Human BCAS2 peptide (14 aa near C-terminus)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5627P
PRSI5627PEA 122 EUR InStock
PRSI5627P
Human BCAS2 peptide (14 aa near C-terminus)
Proteine und Peptide
BCAS2 pepide is used for blocking the activity of BCAS2 antibody.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge