Mouse recombinant darcin (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-264
PRSI90-264EA 448 EUR InStock
PRSI90-264
Mouse recombinant darcin (from E. coli)
Proteine und Peptide
Darcin (also called Mup20) belongs to the family of major urinary proteins (MUPs), a subfamily of proteins found in abundance in the urine. Like all MUPs, Darcin is a member of the lipocalin family of ligand-binding proteins, and preferentially binds the volatile pheromone (S)-2-sec-butyl-4,5-dihydrothiazole (SBT). Darcin is present in the urine of male and acts as a sex pheromone that elicits female sexual attraction to a male's urine scent. Recombinant Darcin shows similar activity compared to natural Darcin.
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge