Human recombinant CD40L multimeric (from CHO cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-289
PRSI90-289EA 620 EUR InStock
PRSI90-289
Human recombinant CD40L multimeric (from CHO cells)
Proteine und Peptide
MultimericCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to CD40L+enhancer combinations. MultimericCD40L™ has been shown to suppress alum-induced IL-1beta release and caspase-1 activation in a dose-, CD40- and time dependent manner, without affecting BMDM (Bone marrow-derived macrophages) viability. It also effectively suppressed the inflammasome function triggered by NLRP3 activators. The secretion of caspase-1 independent inflammatory mediators has been shown to be unaltered or even enhanced.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge