Mouse recombinant Cochlin LCCL domain (from HEK293 cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-314
PRSI90-314EA 620 EUR InStock
PRSI90-314
Mouse recombinant Cochlin LCCL domain (from HEK293 cells)
Proteine und Peptide
Cochlin is an extracellular matrix protein expressed in the cochlea and vestibule of the inner ear. Mutations in the cochlin gene are linked to the autosomal-dominant DFNA9 syndrome, a disorder characterised by hearing loss. Cochlin is also expressed in follicular dendritic cells of the spleen and the lymph nodes. The N-terminus of cochlin contains a LCCL domain also present in Factor C, a serine protease of the horseshoe crab Limulus, involved in activating the coagulation cascade in response to LPS. A recent study showed that in response to inflammation, LPS or bacterial infection, cochlin is cleaved by the proteases aggrecanases and secreted into blood circulation. The secreted cochlin (LCCL domain) seems to have a role in local cytokine production, recruitment of immune effector cells and bacterial clearance. Cochlin is secreted as two forms p8 and p18, whereas p18 being the glycosylated form of p8.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge