Human recombinant NMNAT3 (highly active) (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-356
PRSI90-356EA 448 EUR InStock
PRSI90-356
Human recombinant NMNAT3 (highly active) (from E. coli)
Proteine und Peptide
Mitochondrial NMNAT isoform. Catalyses the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form of nicotinic acid mononucleotide (NAMN) as substrate with the same efficiency. Can use tiazofurin monophosphate as substrate. Can also use GTP and ITP as nucleotide donors. Also catalyses the reverse reaction, i. e. the pyrophosphorolytic cleavage of NAD+. For the pyrophosphorolytic activity, mitochondrial NMNAT isoform can use NAD(+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. Fails to cleave phosphorylated dinucleotides NADP+, NADPH and NAADP+. Protects against axonal degeneration following injury.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge