Recombinant Zmp1 M. tuberculosis (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-361
PRSI90-361EA 773 EUR InStock
PRSI90-361
Recombinant Zmp1 M. tuberculosis (from E. coli)
Proteine und Peptide
M13 endopeptidases are single-pass, transmembrane zinc-metallopeptidases responsible in human for the regulation of the biological activity of many hormones and peptides and are involved in many important processes such as blood pressure regulation (Neprilysin or NEP1), cardiovascular development (Endothelin Converting Enzyme-1 or ECE-12), prevention of haemolytic reaction (KELL) and phosphate homeostasis (PHEX). Zmp1 is a soluble enzyme member of the M13 endopeptidases family that plays a key role in Mycobacterium tuberculosis pathogenicity with a still unknown mechanism. Recently, it was shown to suppress inflammasome activation by inhibiting caspase-1 activation, thus preventing processing of pro-IL-1beta into IL-1beta and the consequent phagosome maturation.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge