Mouse recombinant CD274 non-lytic (from CHO cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-432
PRSI90-432EA 654 EUR InStock
PRSI90-432
Mouse recombinant CD274 non-lytic (from CHO cells)
Proteine und Peptide
Programmed death ligand 1 (PD-L1, B7-H1 or CD274) is a member of the growing B7 family of immune proteins that provide signals for both stimulating and inhibiting T cell activation. CD274 has been identified as one of two ligands for programmed death 1 (PD-1), a member of the CD28 family of immunoreceptors. CD274 is widely expressed in several organs such as heart, skeletal muscle, placenta and lung, and in lower amounts in thymus, spleen, kidney and liver. CD274 expression is upregulated in a small fraction of activated T and B cells and a much larger fraction of activated monocytes. CD274 expression is also induced in dendritic cells and keratinocytes after IFN-gamma stimulation. CD274 expression is also upregulated in a variety of tumour cell lines. Interaction of CD274 with PD-1 results in inhibition of TCR mediated proliferation and cytokine production, suggesting an inhibitory role in regulating immune responses. The CD274 - PD-1 pathway is involved in the negative regulation of some immune responses and may play an important role in the regulation of peripheral tolerance.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge