Human recombinant IL24 (from CHO cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-527
PRSI90-527EA 686 EUR InStock
PRSI90-527
Human recombinant IL24 (from CHO cells)
Proteine und Peptide
IL-24 is a cytokine belonging to the IL-10 family of cytokines that signals through two heterodimeric receptors: IL-20R1/IL-20R2 and IL-22R1/IL-20R2. This interleukin is also known as melanoma differentiation-associated 7 (mda-7) due to its discovery as a tumour suppressing protein. IL-24 appears to control cell survival and proliferation by inducing rapid activation of particular transcription factors called STAT1 and STAT3. It is predominantly released by activated monocytes, macrophages and T helper 2 (Th2) cells and acts on non-haematopoietic tissues such as skin, lung and reproductive tissues. IL-24 performs important roles in wound healing, psoriasis and cancer. IL-24 has antiproliferative properties is inhibiting angiogenesis and may contribute to terminal cell differentiation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge