Human recombinant Carbonic Anhydrase 8 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-095
PRSI91-095EA 469 EUR InStock
PRSI91-095
Human recombinant Carbonic Anhydrase 8 (from E. coli)
Enzyme
Carbonic Anhydrase 8 (CA8) belongs to the alpha-carbonic anhydrase family. Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Because CA8 has some sequence similarity with other known carbonic anhydrase genes, it was firstly called as CA-related protein. Nevertheless, CA8 does not have a carbonic anhydrase catalytic activity. Defects in CA8 are the cause of cerebellar ataxia mental retardation and dysequilibrium syndrome type 3 (CMARQ3), which is a congenital cerebellar ataxia associated with dysarthia, quadrupedal gait and mild mental retardation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge