Human recombinant STUB1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-098
PRSI91-098EA 654 EUR InStock
PRSI91-098
Human recombinant STUB1 (from E. coli)
Proteine und Peptide
E3 Ubiquitin-Protein Ligase CHIP is a cytoplasmic protein. CHIP is highly expressed in skeletal muscle, heart, pancreas, brain and placenta. CHIP interacts with the molecular chaperones Hsc70-Hsp70 and Hsp90 through its TPR domain; lead to in client substrate ubiquitylation and degradation by the proteasome. CHIP targets misfolded chaperone substrates towards proteasomal degradation. CHIP mediates transfer of non-canonical short ubiquitin chains to HSPA8 that have no effect on HSPA8 degradation. CHIP plays a role in base-excision repair: catalyzes polyubiquitination by amplifying the HUWE1/ARF-BP1-dependent monoubiquitination and leading to POLB-degradation by the proteasome. It also may regulate the receptor stability and activity through proteasomal degradation.
  • This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge