Human recombinant Cornulin (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-100
PRSI91-100EA 654 EUR InStock
PRSI91-100
Human recombinant Cornulin (from E. coli)
Proteine und Peptide
Cornulin is a member of the fused gene family of molecular chaperones. Human Cornulin contains N-terminus EF-hand domains and Ca2+ binding domains, and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. Cornulin involves in the mucosal/epithelial immune response and epidermal differentiation. Cornulin is a survival factor that participates in the clonogenicity of squamous esophageal epithelium cell lines, attenuates deoxycholic acid (DCA)-induced apoptotic cell death and release of calcium. When Cornulin is overexpressed in oral squamous carcinoma cell lines, it regulates negatively cell proliferation by the induction of G1 arrest.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge