Human recombinant Cytoglobin (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-107
PRSI91-107EA 654 EUR InStock
PRSI91-107
Human recombinant Cytoglobin (from E. coli)
Proteine und Peptide
Cytoglobin is a ubiquitously globin protein that belongs to the globin family. The highest expressed in heart, stomach, bladder and small intestine. CYGB acts a protector under conditions of oxidative stress. CYGB may be involved in intracellular oxygen storage or transfer, modulates oxygen and nitric oxide metabolism or scavenging free radicals within a cell.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge