Human recombinant FABP3 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-118
PRSI91-118EA 654 EUR InStock
PRSI91-118
Human recombinant FABP3 (from E. coli)
Proteine und Peptide
Fatty Acid Binding Protein 3 (FABP3) is a small cytoplasmic protein (15 kDa) that is released from cardiac myocytes following an ischemic episode. Like the nine other distinct FABPs that have been identified, FABP3 is involved in active fatty acid metabolism where it transports fatty acids from the cell membrane to mitochondria for oxidation. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-types. They form 14-15 kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. The FABP3 gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is also a candidate tumor suppressor gene for human breast cancer. FABP3 is a sensitive biomarker for myocardial infarction and can be detected in the blood within one to three hours of onset of pain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge