Human recombinant NPPB (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-136
PRSI91-136EA 654 EUR InStock
PRSI91-136
Human recombinant NPPB (from E. coli)
Proteine und Peptide
Human Natriuretic peptides B acts as a cardiac hormone; it is associated with many biological actions, such as diuresis, natriuresis, vasorelaxation, which inhibits the secretion of rennin and aldosterone. It acts as a paracrine antifibrotic factor in the heart. Natriuretic peptides B can help restore the body balance of salt and water, improves the heart function. Natriuretic peptides B binds and stimulates the cGMP production of the NPR1 receptor and binds the clearance receptor NPR3.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge