Human recombinant Cytochrome b5A (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-197
PRSI91-197EA 852 EUR InStock
PRSI91-197
Human recombinant Cytochrome b5A (from E. coli)
Proteine und Peptide
Cytochrome b5 (CYB5A) is a membrane bound hemoprotein which function as an electron carrier for several membrane bound oxygenases. CYB5A contains one cytochrome b5 heme-binding domain and has two isoforms produced by alternative splicing. Isoform 1 is a sngle-pass membrane protein. Isoform 2 is located in cytoplasm. The defects in CYB5A can result in type IV hereditary methemoglobinemia.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge