Human recombinant GGACT (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-208
PRSI91-208EA 884 EUR InStock
PRSI91-208
Human recombinant GGACT (from E. coli)
Proteine und Peptide
Gamma-Glutamylaminecyclotransferase is an enzyme that converts gamma-glutamylamines to free amines and 5-oxoproline which belongs to the gamma-glutamylcyclotransferase family. It shows high activity toward gamma-glutamyl-epsilon-lysine, derived from the breakdown of fibrin and contributes to degradation of proteins cross-linked by transglutaminases. It degrades the cross-link between a lysine and a glutamic acid residue from two proteins that have been cross-linked by transglutaminases. This protein adopts the newly identified cyclotransferase fold, observed in Gamma-Glutamylcyclotransferase, an enzyme with activity toward gamma-glutamyl-alpha-amino acids.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge