Human recombinant heat shock beta 11 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-231
PRSI91-231EA 884 EUR InStock
PRSI91-231
Human recombinant heat shock beta 11 (from E. coli)
Proteine und Peptide
Heat Shock Protein beta -11 (HSPB11) is a stress-responsive protein that is required to deal with proteotoxic stresses. HSPB11 is composed of an IFT complex B composed of IFT88, IFT57, TRAF3IP1, IFT52, IFT27, HSPB11 and IFT20 and is detected in placenta. HSPB11 has beeb shown to form oligomeric complexes and prevent the aggregation of in vitro denaturated aldolase and glyceraldehyde-3-phosphate dehydrogenase in accordance with the chaperone model of HSPB1 and HSPB5. HSPB11 overexpression protected against etoposide-induced cell death that correlated with a decreased release of mitochondrial Cytochrome C into the cytosol. Inhibition of HSP90 function completely abrogated the protective effect of HSPB11. This data suggests that at least in the case of HSPB11, interaction with other chaperone machines besides HSPA1A may contribute to functional specificity and cellular functioning.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge