Human recombinant S100 calcium binding P (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-238
PRSI91-238EA 884 EUR InStock
PRSI91-238
Human recombinant S100 calcium binding P (from E. coli)
Proteine und Peptide
Protein S100-P (S100P) belongs to the S100 family of Calcium-Binding Proteins. S100P is 95 amino acids in length and it contains 2 EF-Hand Calcium-Binding Motifs. The EF-Hand Motif binds calcium, which likely alters molecular conformation. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and it is involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. In addition to binding Ca, S100P also binds Zn and Mg, and may play a role in the etiology of prostate cancer.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge