Human recombinant CLIC2 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-246
PRSI91-246EA 884 EUR InStock
PRSI91-246
Human recombinant CLIC2 (from E. coli)
Proteine und Peptide
Chloride Intracellular Channel Protein 2 (CLIC2) is a critical component of all living cells; it regulatescellular traffic of Chloride ion and it can be inserted into membranes anf form chloride ion channels. Membrane insertion seems to be redox-regulated and may occur only under oxydizing conditions, channel activity depends on the pH. CLIC2 is involved in regulating membrane potential and organic solute transport. CLIC2 modulates the activity of RYR2 and inhibits Calcium influx. CLIC2 can be detected in the adult brain, liver, lung, heart, stomach, spleen and testis. It is expressed in fetal liver and adult skeletal muscle. CLIC2 is a potential candidate for one of many diseases linked to Xq28.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge