Human recombinant Malate dehydrogenase (MDH) (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-256
PRSI91-256EA 884 EUR InStock
PRSI91-256
Human recombinant Malate dehydrogenase (MDH) (from E. coli)
Enzyme
Malate Dehydrogenase, Cytoplasmic (MDH1) is an enzyme which belongs to the MDH Type 2 sub-family of LDH/MDH superfamily. MDH1 is involved in the Citric Acid Cycle that catalyses the conversion of Malate into Oxaloacetate (using NAD+) and vice versa. MDH1 should not be confused with Malic Enzyme, which catalyses the conversion of Malate to Pyruvate, producing NADPH. MDH1 also participates in Gluconeogenesis, the synthesis of Glucose from smaller molecules. Pyruvate in the mitochondria is acted upon by Pyruvate Carboxylase to form Pxaloacetate, a Citric Acid Cycle intermediate. In order to transport the Oxaloacetate out of the Mitochondria, Malate Dehydrogenase reduces it to Malate, and it then traverses the inner Mitochondrial membrane. Once in the cytosol, the Malate is oxidised back to Oxaloacetate by MDH1. Finally, Phosphoenol-Pyruvate Carboxy Kinase (PEPCK) converts Oxaloacetate to Phosphoenol Pyruvate.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge