Human recombinant Hydroxyacid Oxidase 1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-259
PRSI91-259EA 884 EUR InStock
PRSI91-259
Human recombinant Hydroxyacid Oxidase 1 (from E. coli)
Enzyme
Hydroxyacid Oxidase 1 (HAO1) is an enzyme that belongs to the FMN-Dependent alpha-Hydroxy Acid Dehydrogenase family. HAO1 contains 1 FMN Hydroxy Ccid Dehydrogenase domain. HAO1 is expressed primarily in the liver and pancreas. This protein has 2-Hydroxyacid Oxidase activity. Most HAO1 is active on the 2-Carbon substrate Glycolate, but it can also be active on 2-Hydroxy fatty acids, with higher activity towards 2-Hydroxy Palmitate and 2-Hydroxy Octanoate.
  • Fusion-Tag: N-Trx, 6 His tag
Fusion-Tag: N-Trx, 6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge