Human recombinant EPT1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-263
PRSI91-263EA 982 EUR InStock
PRSI91-263
Human recombinant EPT1 (from E. coli)
Proteine und Peptide
Ethanolaminephosphotransferase 1 (EPT1) is an enzyme that belongs to the CDP-Alcohol Phosphatidyltransferase Class-I Family. EPT1 is a Selenoprotein, which contains a Selenocysteine (Sec) residue at its active site. The Selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of Selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. EPT1 catalyzes Phosphatidylethanolamine biosynthesis from CDP-Ethanolamine. It plays a central role in the formation and maintenance of vesicular membranes. EPT1 is involved in the formation of Phosphatidylethanolamine via the 'Kennedy' pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge