Human Recombinant GTF2B (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-273
PRSI91-273EA 852 EUR InStock
PRSI91-273
Human Recombinant GTF2B (from E. coli)
Proteine und Peptide
Transcription Initiation Factor IIB (TFIIB) is an essential factor for transcription by RNA Polymerase II. TFIIB localizes to the nucleus where it forms a complex (the DAB complex) with transcription factor IID and IIA. TFIIB plays a role as a bridge between IID, which initially recognizes the promoter sequence, and RNA polymerase II. TFIIB is involved in the selection of transcription start site.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge