Human recombinant Endocan (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-313
PRSI91-313EA 549 EUR InStock
PRSI91-313
Human recombinant Endocan (from cells)
Proteine und Peptide
ESM-1, short from Endothelial cell-specific molecule 1, is expressed in lung, on the vascular capillary network within alveolar walls, and also at lower level in kidney. It is a secretory protein, and can be inducted by TNF and IL1B/interleukin-1beta, but not IL4/interleukin-4. This protein plays great roles in angiogenesis, sprouting, and may have potent implications in lung endothelial cell-leukocyte interactions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge