Human recombinant IGF Binding 4 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-320
PRSI91-320EA 734 EUR InStock
PRSI91-320
Human recombinant IGF Binding 4 (from cells)
Proteine und Peptide
IGFBP-4, whose full name is insulin-like growth factor-binding protein 4, is induced by forskolin and N6, O2’dibutyryl sdenosine 3’, or 5’-cyclic monophosphate. It contains IGFBP N-terminal domain and thyroglobulin type-1 domain, and can bind IGF2. The IGF-binding proteins can prolong the half-life of the IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge