Human recombinant alpha-Galactosidase A (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-323
PRSI91-323EA 734 EUR InStock
PRSI91-323
Human recombinant alpha-Galactosidase A (from cells)
Enzyme
alpha-Galactosidase A is a homodimeric glycoprotein that belongs to the glycosyl hydrolase 27 family. It is a lysosomal enzyme and used as a long-term enzyme replacement therapy in patients with a confirmed diagnosis of Fabry disease. alpha-Galactosidase A can hydrolyze terminal alpha-galactosyl moieties from glycolipids and glycoproteins and catalyze the hydrolysis of melibiose into galactose and glucose. Defects alpha-Galactosidase A are the cause of Fabry disease (FD) which is a rare X-linked sphingolipidosis disease with glycolipid accumulates in many tissues. The disease consists of an inborn error of glycosphingolipid catabolism. FD patients show systemic accumulation of globotriaoslyceramide (Gb3) and related glycosphingolipids in the plasma and cellular lysosomes throughout the body. Patients may show ocular deposits, febrile episodes, and burning pain in the extremities. Death results from renal failure, cardiac or cerebral complications of hypertension or other vascular disease.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge