Human recombinant IFN alpha (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-329
PRSI91-329EA 549 EUR InStock
PRSI91-329
Human recombinant IFN alpha (from cells)
Proteine und Peptide
The Interferon- alpha/ beta Receptor 1 (IFN- alpha/ beta R1) is a receptor which binds Type I Interferons including Interferon- alpha and - beta . It is a cell surface receptor and heteromeric receptor composed of one chain with two subunits referred to as IFNAR1 and IFNAR2. IFN- alpha/ beta R1, in association with IFN- alpha/ beta R2, is required for propagating antiviral signal transduction triggered by IFN- alpha and IFN- beta . IFN- alpha/ beta R1 interacts very weakly or not at all with type 1 interferons and does not stably interact with IFN- alpha/ beta R2. Ligands associate with IFN- alpha/ beta R2, and this complex subsequently forms a stable ternary assembly with IFN- alpha/ beta R1. IFN- alpha/ beta R1 also associates with IFN- gamma R2 even in the absence of IFN- gamma stimulation. Human IFN- alpha/ beta R1 contains a nuclear localization signal in its extracellular domain that is required for receptor translocation to the nucleus following interaction with ligand. Interferon stimulation results in an immunologic response that is especially associated with viruses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge