Human recombinant TREML1 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-358
PRSI91-358EA 528 EUR InStock
PRSI91-358
Human recombinant TREML1 (from cells)
Proteine und Peptide
Triggering Receptor Expressed on Myeloid Cells-Like Protein 1 (TREML1) is a single-pass type I membrane protein. TREML1 precursor contains a 15 amino acid signal peptide, a 147 amino acid extracellular domain with an Ig-like V-type (immunoglobulin-like) domain, and 128 amino acid cytoplasmic domain. It can be expressed exclusively in platelets and megakaryocytes (MKs). It is a cell surface receptor that may play a role in the innate and adaptive immune response. TREML1 Sequestered in cytoplasmic vesicles in resting platelets. TREML1 be transported to the cell surface after stimulation by thrombin. Soluble fragments can be released into the serum by proteolysis. The phosphorylated TREML1 can interact with PTPN6 and PTPN11. TREML1 may participate in maintaining vascular hemostasis and regulating coagulation and inflammation at sites of injury.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge