Human recombinant UPA (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-360
PRSI91-360EA 884 EUR InStock
PRSI91-360
Human recombinant UPA (from cells)
Proteine und Peptide
Recombinant Human Urokinase-Type Plasminogen Activator is a serine protease, which specifically cleaves the zymogen plasminogen to form the active enzyme plasmin. Urokinase-Type Plasminogen Activator is a potent marker of invasion and metastasis in many human cancers associated with breast, colon, stomach, bladder, brain, ovary and endometrium. Human Urokinase-Type Plasminogen Activator is initially synthesized as 431 amino acid precursor with a N-terminal signal peptide residues. The single chain molecule is processed into a disulfide-linked two-chain molecule. There exists two forms A chain, the long A chain contains an EGF-like domain that is responsible for binding of the uPA receptor. The B chain corresponds to the catalytic domain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge