Human recombinant AG-2 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-380
PRSI91-380EA 654 EUR InStock
PRSI91-380
Human recombinant AG-2 (from cells)
Proteine und Peptide
Anterior Gradient 2 (AGR2) is an 18-21 kDa member of the PDI family of enzymes. AGR2 is widely expressed in secretory cells, such as small intestine goblet, prostate epithelium, enteroendocrine cells, and multiple carcinoma cell types. AGR2 forms transient disulfide linkages with molecules destined for secretion, possibly aiding protein folding. Expression of AGR2 shows a positive correlation with expression of estrogen receptor in breast carcinoma and a negative correlation with expression of EGF receptor. Mature human AGR2 is 155 amino acids (aa) in length (aa 21 - 175). Cys81 is presumed to participate in intermolecular bond formation. Over aa 21 - 175, human AGR2 shares 94% aa identity with mouse AGR2.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge