Human recombinant ASAM (from Cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-383
PRSI91-383EA 549 EUR InStock
PRSI91-383
Human recombinant ASAM (from Cells)
Proteine und Peptide
Adipocyte Adhesion Molecule (ASAM) is a type I transmembrane protein and member of the CTX family within the immunoglobulin superfamily. ASAM may be involved in the cell-cell adhesion, play an important role in adipocyte differentiation and development of obesity. ASAM can be expressed in the skeletal, heart, colon, spleen, muscle, lung and kidney with high level, and in the peripheral blood leukocytes and liver with low level. The extracellular region of ASAM consists two potential N-linked glycosylation sites, and two immunoglobulin domains, one V-type and one C2-type.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge