Human recombinant TrkB (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-458
PRSI91-458EA 377 EUR InStock
PRSI91-458
Human recombinant TrkB (from cells)
Proteine und Peptide
The TRK Family of Tyrosine Kinase Receptor consists of 3 members: TrkA, TrkB and TrkC. The three TRK family proteins have different ligand specificities. They connect to different neurotrophins, including NGF, BDNF, NT-3NT-4/5. TRKA binds NGF, TRKB binds BDNF and NT-3, TRKC binds NT-4/5. At the protein sequence level, human and rat TRKB have greater than 90% sequence identity and the proteins exbihit cross-species activity. TRKB is primarily expressed in the nervous system and it also expression in a wide variety of tissues with low levels.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge