Human recombinant TEM7 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-479
PRSI91-479EA 654 EUR InStock
PRSI91-479
Human recombinant TEM7 (from cells)
Proteine und Peptide
Plexin Domain-Containing Protein 1 (PLXDC1) is a single-pass type I membrane protein that belongs to the plexin family. Secreted PLXDC1 is localized predominantly at the tight junctions of vascular endothelial cells and to a lesser extent at the luminal surface of vascular endothelial cells. PLXDC1 is expressed in fibrovascular membrane with increased expression in individuals with proliferative diabetic retinopathy. It can detect in endothelial cells from colorectal cancer, and in endothelial cells from primary cancers of the lung, liver, pancreas, breast and brain. PLXDC1 interacts with NID1 and may also interact with CTTN. It plays a important role in endothelial cell capillary morphogenesis, the proliferation and maintenance of neovascular endothelial cells in the formation of fibrovascular membranes (FVMs).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge