Human recombinant CLEC4E (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-525
PRSI91-525EA 654 EUR InStock
PRSI91-525
Human recombinant CLEC4E (from cells)
Proteine und Peptide
C-Type Lectin Domain Family 4 Member E (CLEC4E) is a 219 amino acid single-pass type II membrane protein that contains one C-type Lectin domain. It is expressed in monocytes, CLEC4E functions as a downstream target of C/EBP beta and is thought to play a role in the inflammatory response, possibly via transcriptional control of C/EBP beta . CLEC4E may play a role in the response to inflammatory stimuli in peritoneal macrophages and may be involved in immune surveillance processes under transcriptional control of CEBPB. Human CLEC4E shares 67% sequence identity with its mouse counterpart, suggesting a similar function between species. CLEC-4E exists as multiple alternatively spliced isoforms that are encoded by a gene which maps to a natural killer gene complex region on human chromosome 12.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge