Human recombinant ST8SIA1 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-569
PRSI91-569EA 852 EUR InStock
PRSI91-569
Human recombinant ST8SIA1 (from cells)
Proteine und Peptide
alpha-N-Acetylneuraminide alpha-2,8-Sialyltransferase (ST8SIA1) belongs to the glycosyltransferase 29 family. ST8SIA1 is a sialytransferase that catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce GD3 and GT3. ST8SIA1 is highly expressed in melanoma cell lines, adult and fetal brain, low expressed in adult and fetal lung. ST8SIA1 may act as a type II transmembrane protein with a short N-termianl cytoplasmic domain and a single-pass transmembrane domain folowed by an enzymatic domain in the lument of the Golgi apparatus.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge