Human recombinant S100 calcium binding A12 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-588
PRSI91-588EA 588 EUR InStock
PRSI91-588
Human recombinant S100 calcium binding A12 (from E. coli)
Proteine und Peptide
Human EN-RAGE belongs to the S100/calgranulin superfamily. There are at least 21 different S100 proteins and the protein is 100% soluble in ammonium sulfate at neutral pH. S100 proteins play a role in regulation of protein phosphorylation, transcription factors, the dynamics of cytoskeleton constituents, enzyme activities, cell growth and differentiation, and the inflammatory response.Human EN-RAGE is characterized by two EF-hand calcium-binding motifs, zinc- and copper-binding protein.S100A12 is a disulfide-linked homodimer and the interface between the two subunits is composed mostly of hydrophobic residues.Its proinflammatory activity involves recruitment of leukocytes, promotion of cytokine and chemokine production, and regulation of leukocyte adhesion and migration. EN-RAGE acts as an alarmin or a danger associated molecular pattern (DAMP) molecule and stimulates innate immune cells via binding to receptor for advanced glycation endproducts (AGER). Binding to AGER activates the MAP-kinase and NF-kappa-B signaling pathways leading to production of proinflammatory cytokines and up-regulation of cell adhesion molecules ICAM1 and VCAM1. It also acts as a monocyte and mast cell chemoattractant. Moreover, it can stimulate mast cell degranulation and activation which generates chemokines, histamine and cytokines inducing further leukocyte recruitment to the sites of inflammation. It can inhibit the activity of matrix metalloproteinases; MMP2, MMP3 and MMP9 by chelating Zn2+from their active sites.
  • This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge