Human recombinant roundabout homolog 4 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-612
PRSI91-612EA 588 EUR InStock
PRSI91-612
Human recombinant roundabout homolog 4 (from cells)
Proteine und Peptide
Roundabout homolog 4(ROBO4) contains 2 fibronectin type-III domains, 2 Ig-like C2-type (immunoglobulin-like) domains and belongs to the immunoglobulin superfamily.The protein is a receptor for slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. ROBO4 may mediate the inhibition of primary endothelial cell migration by Slit proteins.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge