Human recombinant VAMPB (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-620
PRSI91-620EA 588 EUR InStock
PRSI91-620
Human recombinant VAMPB (from E. coli)
Proteine und Peptide
vesicle-associated membrane protein-associated protein B/C (VAPB) is presents in mitochondria-associated membranes that are endoplasmic reticulum membrane regions closely apposed to the outer mitochondrial membrane. It can form homodimer, and heterodimer with VAPA. It also interacts with VAMP1, VAMP2, HCV NS5A, NS5B, ZFYVE27 and RMDN3. VAPB participates in the endoplasmic reticulum unfolded protein response (UPR) by inducing ERN1/IRE1 activity. It is involved in cellular calcium homeostasis regulation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge