Human recombinant Carbonic Anhydrase 11 (from Cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-650
PRSI91-650EA 884 EUR InStock
PRSI91-650
Human recombinant Carbonic Anhydrase 11 (from Cells)
Enzyme
Carbonic Anhydrase-Related Protein 11 (CA11) is a secreted protein member of the alpha-carbonic anhydrase family. Carbonic Anhydrases (CAs) are a large family of zinc metalloenzymes that catalyse the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. CA11 is expressed abundantly in the brain with moderate expression also present in spinal cord and thyroid. CA11 may play a general role in the central nervous system.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge